X-ray and metal detectable bottles, wide neck
Fournisseur: Sampling Systems
M1047B-250
SAMPM1047B-250EA 30.1 EUR InStock
SAMPM1047B-250
X-ray and metal detectable bottles, wide neck
Flacons Flacons en plastique
X-ray and metal detectable bottles are perfect for both laboratory and production work. They are available in a handy 250 ml size with a wide neck. The bright blue colour makes the bottles easy to spot if they are left in the product.
  • Robust PP construction. PP gives excellent chemical resistance to many products
  • 250 ml volume
  • Full batch traceability
  • Non sterile
To ensure the cleanliness of the bottles, they are manufactured and packed in a cleanroom. The robust nature of these bottles means that they can give many years of service or for more demanding applications they can be used as a single use disposable product straight from the bag.
Certifications: These PP bottles conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Conditionnement: Supplied individually bagged.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité