Blood collecting systems, S-Monovette® CTAD
Fournisseur: SARSTEDT
04.1909.001
SARS04.1909.001EA 345.8 EUR InStock
SARS04.1909.001
Blood collecting systems, S-Monovette® CTAD
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
S-Monovette® is a blood collection system that combines two blood collection techniques – aspiration and vacuum. The S-Monovette® is suitable for all vein conditions and achieves an optimal sample quality, thereby producing the best results. The aspiration technique is a gentle technique for routine blood collection. Using the vacuum technique, a "fresh" vacuum is always available.
The S-Monovette® CTAD inhibits thrombocyte activation for a period of at least 4 hours. As a result, there is a similar delay in the release of proteins such as platelet factor 4 (PF4), β-thromoglobulin (βTG)More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité