Sterility test system, Sterisart® NF
Fournisseur: Sartorius
SART16475-GSDEA 542 EUR InStock
SART16475-GSD SART16468-GBD SART16420 SART16467-GSD SART16469-GSD SART16466-GSD SART16476-GBD SART16478-GBD SART16475-GBD SART16467-GBD SART16470-GBD SART16469-GBD SART16466-GBD SART16476-GSD SART16477-GBD
Sterility test system, Sterisart® NF
Filtres Unités à membrane filtrante
The Sterisart® NF System is a completely disposable technology based on the membrane filtration method, for the sterility testing of pharmaceutical products. It makes aseptic sampling easy, during or after incubation which is required for dilution or rapid detection, or for the supplement of additives for inactivation of antibiotics.
  • Sterisart NF alpha, dual-needle metal spike for closed containers
  • Sharp Metal needle for easy penetration
  • High retention of microbes
  • Low adsorption
  • Huge variety of adapters and more than 20 different unit types
  • Gas-impermeable packaging for protection against sterilant
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité