Octet® Anti-penta-HIS (HIS1K) biosensors
Fournisseur: Sartorius
18-5121
SART18-5121EA 4080 EUR InStock
SART18-5121
Octet® Anti-penta-HIS (HIS1K) biosensors
Tests Essais protéiniques
For label-free quantitation and kinetic analysis of his-tagged proteins.
  • High specificity quantitation of his-tagged proteins
  • Easy capture of his-tagged proteins for kinetic analysis with interacting protein analyte
  • High specificity and High affinity towards polyhistidine tag
  • Allows rapid analysis in purified or crude samples
The polyhistidine tag, commonly known as His-tag, is fused to recombinant proteins as a means of facilitating detection and purification. The dip and read anti-penta-HIS (HIS1K) biosensor consists of high affinity, high specificity penta-his antibody from QIAGEN pre-immobilized on a sartorius fibre optic biosensor.
In conjunction with the Octet® systems, the HIS1K biosensor provides a rapid and label-free method for his-tagged protein quantitation and kinetic analysis. The high specificity of the antibody-based biosensor enables the direct capture and quantitation of his-tagged proteins in crude lysates, column eluates, cell lysates and cell culture supernatants, serving as an alternative to traditional time consuming analytical methods such as HPLC and ELISA.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité