Filter cartridges, Sartofluor® GA
Fournisseur: Sartorius
5182507T2GA 5181407T7-B 5182507T3-GA
SARP5182507T2GAEA 1060 EUR InStock
SARP5182507T2GA SART5181407T7-B SARP5182507T3-GA
Filter cartridges, Sartofluor® GA
Filtres Cartouches filtrantes
Sartofluor® GA filter cartridges, manufactured with permanently hydrophobic PTFE membranes, are specially designed for sterile venting and gas applications. Due to their permanent hydrophobicity, they offer the highest process security, even with high volume gas streams, extreme humidity and stringent in-line steam sterilisations.
  • Withstand high differential pressures in either the forward or reverse direction of flow
Membrane: Hydrophobic PTFE; support layers: PP; end cap, core and cage: PP; O-rings: silicone (EPDM or fluoroelastomer)
Maximum differential pressure: five bar at 20 °C; two bar at 80 °C
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité