Filtration capsules, Sartofluor®
Fournisseur: Sartorius
5185307T9-SS-A 5181307T4-OO-B 5185307T7-OO-A 5185307T9-OO-A 5185307T7-SS-A 5185307T9-SO-A 5185307T7-SO-A 5181307T4-SO-B
SARP5185307T9-SS-AEA 984 EUR InStock
SARP5185307T9-SS-A SARP5181307T4-OO-B SARP5185307T7-OO-A SARP5185307T9-OO-A SARP5185307T7-SS-A SART5185307T9-SO-A SARP5185307T7-SO-A SARP5181307T4-SO-B
Filtration capsules, Sartofluor®
Filtres Capsules de filtration
Sartofluor® MidiCaps® and Capsules are self contained, ready to use, sterile filter units for sterilising grade filtration in the pharmaceutical and biotechnology industry. Their unique hydrophobic PTFE membrane is ideally suited for particle removal and sterilising grade filtration of gases and for filtration of highly aggressive liquids like solvents, acids and bases.
  • Easy to use
  • Flexible and cost saving
  • Excellent flow rates for gases and liquids at low differential pressure
PTFE membrane; support fleece, core and end caps: PP; O-rings: EPDM; housing: PP; filling bell: PC
Sartofluor® MidiCaps and capsules are designed, developed and manufactured in accordance with a ISO 9001 certified Quality Management System. A Validation Guide is available for compliance with regulatory requirements.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité