Paper boards for filtration and absorption of liquids
Fournisseur: Sartorius
FT-2-343580580
SARTFT-2-343580580EA 183 EUR InStock
SARTFT-2-343580580
Paper boards for filtration and absorption of liquids
Filtres Papiers filtre Papiers filtres qualitatifs
These paper boards are used for the filtration of cooking and transformer oils, galvanic baths and as base paper for further impregnation with certain reagents. They are made of refined pulp or cotton linters.
  • Smooth
  • Supplied in sheets, discs and as well as customer specific cuts
Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité