Seed testing papers, pleated paper
Fournisseur: Sartorius
FT200353200011
SARTFT200353200011EA 2290 EUR InStock
SARTFT200353200011
Seed testing papers, pleated paper
Filtres Papiers filtre Papiers filtres qualitatifs
These papers satisfy the requirements for the determination of germination capability according to ISTA (International Seed Testing Association) and are ideal for ensuring optimal moisture content for the most diverse types of seeds and germination forms.
  • pH ranges between 6,0 and 7,5
  • Wet-strengthened
  • Special structure prevents fine seed roots from growing through the paper
  • 50 double folds that are 20 mm in depth
The pleated paper is placed in a box; the seeds are distributed among the folds of the pleated paper and covered with a wrapping strip to keep the seeds moist.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité