Real-Time PCR kit for detection of mycoplasma, Microsart® RESEARCH Mycoplasma
Fournisseur: Sartorius
SMB95-1005 SMB95-1006
SARTSMB95-1005EA 346 EUR InStock
SARTSMB95-1005 SARTSMB95-1006
Real-Time PCR kit for detection of mycoplasma, Microsart® RESEARCH Mycoplasma
Tests microbiologiques Tests PCR
The Microsart® RESEARCH Mycoplasma real-time PCR kit is especially designed for fast and reliable Mycoplasma contamination control in cell cultures, cell culture supernatants and media most applicable in research and development, e.g. biotech and biopharmaceutical research and development, universities and governmental research groups or for in-process control during biopharmaceutical production processes.
  • Fast results in less than 3 hours
  • Easy to use
  • Reliable screening with a small sample volume
The 2 µl sample material can be added directly to the PCR reaction tube without the need for any sample preparation. Nevertheless standard DNA isolation procedures can be done optional prior to the DNA amplification. The DNA can be amplified with the help of any qPCR cycler that is able to detect the dyes FAM ™and ROX™ and the evaluation can be performed with the standard cycler software. The use of TaqMan® probes adds specificity to the PCR detection system. Due to the high specificity no melting curve analysis is needed after the amplification steps.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité