Antimicrobial foam soap, Tork
Fournisseur: Essity
520800
SCAA520800EA 140 EUR InStock
SCAA520800
Antimicrobial foam soap, Tork
Soin de la peau Savons et soins nettoyants
Antimicrobial foam soap made of low-alcohol formula is very effective against bacteria, viruses and yeasts. It combines normal hand cleaning with disinfection on one procedure. In addition, no perfume or dyes are incorporated and allergic reactions are thus reduced.
Look: white foam
Color: Transparent
Smell: Alchohol Smell
Certifications: Certified with EN 1499.
Attention: Use biocides safely. Always read the label and product information before use.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité