Griffin beakers, low form, double scale, borosilicate glass, KIMAX®, KIMBLE®
Fournisseur: DWK Life Sciences
SP14000-1500
SCERSP14000-1500EA 398 EUR InStock
SCERSP14000-1500
Griffin beakers, low form, double scale, borosilicate glass, KIMAX®, KIMBLE®
Béchers
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Suitable for use on a hotplate
  • Durable matte finish marking area
  • Easy-to-read white graduated scale
Low form Griffin glass beakers offer excellent mechanical strength and durability, while providing high resistance to chemical attack and thermal shock.
Certifications: Designed from ASTM Specification E960, Type I requirements.
Informations de commande: Starter pack include each one beaker 50/100/250/600 and 1000 ml.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité