Pycnometer, Le Chatelier, class A, serialised, KIMAX®
Fournisseur: DWK Life Sciences
SP15115-24
SCERSP15115-24EA 566 EUR InStock
SCERSP15115-24
Pycnometer, Le Chatelier, class A, serialised, KIMAX®
Flacons de gravité spécifique
KIMAX® bottle is used to determine the specific gravity of cement, sand, and other fine materials (ASTM C188).
  • Designed for use with a 64 g sample
  • Graduated from 18 to 24 ml in 0,1 ml subdivisions
  • All markings are durable white enamel
  • Supplied with serial number
Body holds approximately 250 ml and the smaller neck bulb holds 17 ml. The neck is graduated from 0 to 1,0 ml and has two extra 0,1 ml lines at both above 1,0 ml and below 0,0 ml.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certifications: Designed from ASTM Specification E694 and complies with C188.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité