Chromatography columns, Flex-Column®, KIMBLE®
Fournisseur: DWK Life Sciences
420400-2510 420401-2510 SP420401-1050
KIML420400-2510EA 0 EUR InStock
KIML420400-2510 KIML420401-2510 SCERSP420401-1050
Chromatography columns, Flex-Column®, KIMBLE®
Colonnes pour chromatographie
These economical columns are constructed with borosilicate glass barrels, molded PP end caps, and PE bed supports. They also feature Luer lock inlet and outlet fittings for better connections.
PP packing reservoir funnels are for use in initial column packing or as small buffer reservoirs. Glass packing reservoirs are ideal for packing gel slurries into columns or acting as a buffer reservoir in gravity flow chromatography. Glass reservoirs include a PP upper cap with male Luer lock fitting.
Flow adapters allow easy adjustment of column bed volume and offer improved column performance and chromatogram resolution. Flow adapters are constructed with a PTFE eluant cannula, a Delrin® body, a Viton® O-ring seal, and a 20 µm PE bed support. Each flow adapter includes one replacment bed support; additional replacements are also available.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité