Centrifuge tubes, oil, graduated, KIMAX®
Fournisseur: DWK Life Sciences
SP45240-100
SCERSP45240-100EA 440 EUR InStock
SCERSP45240-100
Centrifuge tubes, oil, graduated, KIMAX®
Tubes Tubes à centrifuger
Borosilicate glass KIMAX®, graduated, snap cap finish.
  • Scale and legend of durable white ceramic enamel
  • Top is tooled to accept a snap cap
It is used in the determination of water and sediment in crude mineral oils, fuel oils and other petroleum products (D1796 and MPMS 10.4 standards), volume of precipitate formed by centrifuging definite quantities of steam cylinder stocks and black oils and other lubricating oils (ASTM D91 and D128), testing for acidity of distillation residues or hydrocarbon liquids of gasoline or petroleum solvents (ASTM D1093).
Tubes are graduated upward to 0,5 ml in divisions of 0,05 ml; from 0,5 ml to 2 ml in divisions of 0,1 ml; from 2 ml to 3 ml in divisions of 0,2 ml; from 3 ml to 5 ml in divisions of 0,5 ml; from 5 ml to 10 ml in divisions of 1 ml; from 10 ml to 25 ml in divisions of 5 ml and at 25 ml to 100 ml in divisions of 25 ml.
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité