EPA vials with PP cap
Fournisseur: DWK Life Sciences
SP60961C-4
SCERSP60961C-4EA 287 EUR InStock
SCERSP60961C-4
EPA vials with PP cap
Fioles
These clear screw thread EPA vials are used for the collection and storage of water samples.
  • Not pre-cleaned
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certifications: These vials comply with the guidelines for establishing test procedures for the analysis of pollutants (Ref. EPA Methods 601, 602, 603 and 604).
Conditionnement: Vials are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and are placedMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité