Scintillation vials, Solvent-Saver™, KIMBLE®
Fournisseur: DWK Life Sciences
SP74503-7
SCERSP74503-7EA 711 EUR InStock
SCERSP74503-7
Scintillation vials, Solvent-Saver™, KIMBLE®
Fioles
Vials are manufactured from 33 expansion, low extractable borosilicate glass. These glass Solvent-Saver™ vials are dimensionally smaller than conventional scintillation vials, permitting a reduction in the amount of solvent required.
  • Trays are shrink-wrapped to preserve cleanliness
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Conditionnement: Vials are packaged in cellular trays of 200, with 5 trays per case. Closures are packed in PE bags of 200 pieces in a separate tray of 1000 pieces.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité