Multi-position magnetic stirrers, MultiMagStir Genie®
Fournisseur: SCIENTIFIC INDUSTRIES ELMIS
SI-3002 SI-3003
SCIISI-3002EA 1530 EUR InStock
SCIISI-3002 SCIISI-3003
Multi-position magnetic stirrers, MultiMagStir Genie®
Agitateurs Agitateurs magnétiques multipostes
The programmable revolutionary 4-position MultiMagStir Genie provides low to high speed stirring and editable memory profiles.
  • Three power levels for various sample viscosities
  • Manual mode provides stir, stir revere, and time functions
  • Timer: 1 second to 99 hours
  • Memory up to 4 user-defined profiles
  • With selectable ramp rate of 5 to 300 min−1/sec
Microprocessor controls offer precision and reproducibility without internal heat build-up. The stirrer incorporates a feature to program user profiles which is ideal for repetitive protocols or for use by different users. Maximum load capacity is 10 kg.
Certifications: ETL listed.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité