ROTO-ROCK GENIE Rotator Rockers
Fournisseur: SCIENTIFIC INDUSTRIES ELMIS
SI-R102
SCIISI-R102EA 1870 EUR InStock
SCIISI-R102
ROTO-ROCK GENIE Rotator Rockers
Agitateurs Agitateurs rotatifs
Programmable combination Rotator/Rocker with digital touch screen, featuring magnetic platform and clip plate system.
  • Intuitive touch screen control provides ease when changing program settings
  • Setting the tilt angle and speed control provides thorough mixing while eliminating spills when using open containers in rocking mode
  • Two-sided magnetic platform holds virtually any type of tube or sample vessel when using GENIE clip plates (clip plates sold separately)
  • Programming modes include rock, rotate, or timed auto-switching between the two
  • Compact design saves valuable bench space in the laboratory
  • Designed for use in cold rooms, incubators (non-CO₂), or at ambient temperature
  • Includes two year warranty
Programable all-in-one benchtop GENIE Rocker/Rotator, or cycle from rock to rotate. Includes magnetic platform and non-slip rocking yray. Accessories sold separately. The GENIE clip plate system is designed to handle a variety of vessels from PCR tubes, vials, bottles, and sealed bags. Digital set/controls for RPM, total time, and cycle time. The unit also features a pause/resume button, allowing the operator to add reagent to a sample during process.
Informations sur les accessoires: Various clip plate accessories available for magnetic platform.
Informations de commande: Supplied with magnetic platform and rocking tray. Excludes clip plate accessories required to attach samples to magnetic platform for rotating. No special handling required.
Conditionnement: Package includes unit plus rocking tray combined.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité