Microbial testing swabs, Promedia
Fournisseur: HYSERVE GMBH & CO. KG
2200001
SCIL2200001EA 114 EUR InStock
SCIL2200001
Microbial testing swabs, Promedia
Écouvillons Écouvillons à usage clinique
This Swab System is designed for sampling on surfaces in the production environment. It consists of a sterile swab in a tube with 10 ml sterile PBS buffer for sampling, transport, and subsequent quantitative detection of present microorganisms in combination with Compact Dry and Easy Plate culture medium plates.
Using the swab stick, a surface is sampled. This sample is homogenously mixed in the buffer medium by vigorously shaking the tube. Subsequently, 1ml of the liquid is transferred onto a culture plate using the outlet at the bottom of the tube. The inoculated culture plates are incubated according to their specification and evaluated by counting the number of specific colonies.
Information de livraison: Kit components: Transport tube with outlet cap, swab stick attached to the cap, PBS buffer (10 ml).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité