22061978
22061977
22061976
22061975
22061974
22061973
SCIL22061978EA
240
EUR
InStock
SCIL22061978
SCIL22061977
SCIL22061976
SCIL22061975
SCIL22061974
SCIL22061973
Kikkoman Easy Plates - Film Culture Media for Microbiological Testing
Tests microbiologiques
Tests sur les micro-organismes
Easy Plate are ready-to-use culture media prepared in an easy-to-use, stackable film plate format that saves space, reduces waste, eliminates preparation time and labor, and streamlines your test procedures.
- Ready and easy to use: Place sample suspension onto the plate and start incubation
- No special tools or spreaders are required due to a sterile ring around the plate that avoids loss of sample
- Reduced incubation space and waste in comparison with standard Petri dishes
- Easily distinguishable brightly colored colonies make it is easy to count colonies
The hydrophobic ring around the plate ensures sample suspension spreads evenly across the plate surface without the need for a spreader.
Certifications: AOAC certified: Independent third-party review and certification for test method performance.
Informations sur les accessoires: Shelf life: 18 months after manufacturing date (12 months for Easy Plate SA)
Information de livraison: 1 box with 100 plates.
Attention: Store at temperatures between 2 and 8 °C.