BioDrop µLite+ Microvolume Spectrophotometer
Fournisseur: BIOCHROM (SCIE-PLAS LTD)
80-3006-55
SCLI80-3006-55EA 11810 EUR InStock
SCLI80-3006-55
BioDrop µLite+ Microvolume Spectrophotometer
Spectrophotomètres et Photomètres Biophotomètres
BioDrop's unique sample port is dedicated to microvolume measurement. The port is easy to use: simply pipette as little as 0,5 μl and measure. Cleaning the port is easy too, just wipe with lint-free tissue to reduce sample carryover to undetectable amounts.
  • The in-built sample port uses no moving parts
  • Instrument provides excellent reproducibility without the need to recondition or calibrate
  • Highly accurate with the pathlength of the port specified to ±5 μm
  • Export data via USB or link to PC
Easy to use onboard analysis with pre-programmed applications or users designed protocols.
Informations de commande: BioDrop Resolution CFR software is available if needed for full 12 CFR Part 11 compliance. IQ/OQ available upon request.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité