Fibre optic ring lights
Fournisseur: SCHOTT AG Lighting and imaging
157066 157081 157086
SCOC157066EA 688 EUR InStock
SCOC157066 SCOC157081 SCOC157086
Fibre optic ring lights
Éclairages Éclairages annulaires
Fibre optic ring lights are available as six point, annular, or darkfield versions with specific advantages. For all ring lights, adapters are available for a variety of objective series. Ring lights are used to provide intense, uniform, and shadow-free illumination.
  • Mounts directly to the objective using a thumb screw
  • Housing made of rugged, black, anodised aluminium
  • Fibre bundle protected by flexible metal PVC sheathing
  • High white light transmission thanks to the use of SCHOTT PURAVIS ® fibres
Annular ring lights offer absolute shadow-free illumination. Six point ring lights are a cost effective alternative to annular ring lights. Darkfield ring lights offer low angle illumination for shiny, reflective surfaces.
Informations de commande: Cold light source must be ordered separately.
Information de livraison: Supplied without cold light source.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité