DURAN® protect Laboratory Bottle, Amber
Fournisseur: DWK Life Sciences
218068687
SCOT218068687EA 948 EUR InStock
SCOT218068687
DURAN® protect Laboratory Bottle, Amber
Flacons Flacons en verre
DURAN® protect Laboratory Bottle, amber, PU plastic safety coated, with thread DIN 168-1, USP <660> (Spectral Transmission) and EP (3.2.1), graduated.
  • Manufactured from ambered borosilicate glass 3.3 with a polyurethane coating
  • The polyurethane PU coating provides scratch protection
  • USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 and 450 nm
  • The only external application of the amber colour and PU coating ensures that internal surfaces have standard DURAN® glass properties
  • Easy-to-read scale with graduation marks to estimate the content
Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders.
Informations de commande: Supplied with push-on protective cover. The applicable screw caps and pouring rings must be ordered separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité