Screw caps, YOUTILITY™ GL 45, DURAN®
Fournisseur: DWK Life Sciences
292292827
SCOT292292827EA 44.2 EUR InStock
SCOT292292827
Screw caps, YOUTILITY™ GL 45, DURAN®
Capuchons Bouchons pour flacons
The ergonomic-shaped YOUTILITY™ GL 45 screw cap features a liner less design compatible with both DURAN® original GL 45 bottles and YOUTILITY™ bottles. The cap is moulded from biocomptatible materials making it suitable for use in pharmaceutical manufacturing.
  • Certificate of Conformance (CoC)
  • Ergonomic shape with a comfortable grip
  • Autoclavable at 121 °C (20 minutes) or 134 °C (three minutes)
  • Max. usage temperature 140 °C
  • Inverted cone shape for a safer grip
The white YOUTILITY™ caps are retrace coded for traceability. The retrace code can be used to download a lot specific certificate of conformance. The white YOUTILITY™ GL 45 cap is used in the manufacture of pharmaceutical drugs, the storage and transport of APIs, bulk drug intermediates, ingredients and media.
Certifications: Polypropylene and masterbatch conform to USP <88> 121 °C Class VI. Retrace code certificate is available for download.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité