Bottle base protector, for 5 L bottles, DURAN®
Fournisseur: DWK Life Sciences
292807358
SCOT292807358EA 33.9 EUR InStock
SCOT292807358
Bottle base protector, for 5 L bottles, DURAN®
Portoirs Portoirs pour flacons
Protects the base of 5 L bottles from damage during transport around the laboratory or production facility.
  • Silicone support base protects 5 L glass bottle base from impact during lifting or handling
  • Autoclavable
Ideal for users who have to handle large 5 litre GL 45 or GLS 80 glass laboratory bottles. One of the risks during transportation is accidental impact, and for this reason, a shock absorbing silicone rubber component protects the base of the bottle. The base can be autoclaved, and washed in a lab washer for reuse.
Application: Safe transport of laboratory bottles, for use with 5 L DURAN® bottles.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité