Screw caps for laboratory bottles, with thermocouple holder
Fournisseur: DWK Life Sciences
299922802
SCOT299922802EA 197 EUR InStock
SCOT299922802
Screw caps for laboratory bottles, with thermocouple holder
Capuchons Bouchons pour flacons
The DURAN® screw cap GL 45 thermocouple holder consists of a holder fitted into a blue DURAN® polypropylene GL 45 cap that accepts up to three separate thermocouple wires used in autoclaves or sterilisers.
  • For use in laboratory autoclaves and sterilisers
  • Maximum temperature 140 °C
  • Autoclavable: 121 °C
A particular issue when using thermocouples is that their very thin connection wires have a tendency to curl, making it difficult to maintain the tip (where the temperature is measured) in the correct position within the bottle during the entire sterilising cycle. To overcome this problem, the DURAN® screw cap GL 45 thermocouple holder has a hollow PTFE tube to keep the wires straight.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité