Reusable half masks, Profile2
Fournisseur: SCOTT SAFETY
5032271
SCTS5032271EA 0 EUR InStock
SCTS5032271
Reusable half masks, Profile2
Respirators Masques réutilisables Demi-masques
Profile² is an advanced twin filter half mask with odourless, lightweight TPE facepiece that combines comfort, protection and low through life costs with exceptionally low breathing resistance. The easy fit bayonet filter mechanism enables fast, secure first time connection.
  • Large diameter valves reduce wearer fatigue
  • Comfortable head harness and neck buckle provide full support
  • Latex and silicone-free to ensure no allergic skin reaction or irritation
  • Low profile and swept back filters provide balance and unobstructed field of vision
  • Spark, splash and contaminant guards for filters and exhale valve
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité