Built-in signalbox and switch amplifiers, SymLine®
Fournisseur: S.C.A.T.
108219 106590 106658 106678
SCTV108219EA 82.9 EUR InStock
SCTV108219 SCTV106590 SCTV106658 SCTV106678
Built-in signalbox and switch amplifiers, SymLine®
Capteurs de niveau
Monitor two filling levels at once. Perfect integration into lab furniture with the Built-In signalbox and switch amplifier.
  • Monitor two filling levels at once
  • Touchpad control
  • Level control through status LEDs
  • Optical and acoustic alarm signals
  • Compatible with switch amplifier
LEDs and acoustic buzzer provide warning in due time before the filling level of collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via the integrated interface. According to ATEX Directives, Ex-sensors have to be installed in conjunction with a switch amplifier when used in explosion-prone areas (Ex-zones). Signalboxes and switch amplifier must be placed outside of the Ex-zone.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité