SymLine Automatic Switching for Container
Fournisseur: S.C.A.T.
108334-T 107167 107165 106741 106732 107168 107166 399204 108335-T 108336-T 117111 307338 307380 307448 307970 307971 318958 399203 106751 106752 106733 106735
SCTV108334-TEA 228 EUR InStock
SCTV108334-T SCTV107167 SCTV107165 SCTV106741 SCTV106732 SCTV107168 SCTV107166 SCTV399204 SCTV108335-T SCTV108336-T SCTV117111 SCTV307338 SCTV307380 SCTV307448 SCTV307970 SCTV307971 SCTV318958 SCTV399203 SCTV106751 SCTV106752 SCTV106733 SCTV106735
SymLine Automatic Switching for Container
Matériel de chromatographie Chromatographie, Pièces de rechange
The ready-to-use automatic switching sets prevent the overflowing of disposal containers and the associated leakage of hazardous, toxic, or explosive liquid waste.
  • Coordinated system for monitoring the safe filling of disposal containers
  • For connection to two disposal containers (not included in the set)
  • Set consists of Built-in Signalbox2, SwitchBox, ball valve, and two signal cables
  • Optionally available as an ATEX version with an additional switch amplifier for two channels
Expandable sets for safe fill level monitoring during disposal in containers.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité