Flat canisters
Fournisseur: S.C.A.T.
107713
SCTV107713EA 31.7 EUR InStock
SCTV107713
Flat canisters
Bidons de sécurité
Due to their flat design, SCAT flat canisters have been specially developed for use in confined spaces such as cupboards, shelves, drawers or refrigerators, as they are structurally low in height.
  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in laboratories
  • Available with B63 or S70/71 threads and can be adapted to many other thread sizes by means of an adapter
  • Available either as compact 8 L canister or as more voluminous 20 L canister
SCAT flat canisters are an ideal way of not having to dispense with safe storage even when space is limited, as they can be integrated into laboratory furniture to save space.
SCAT flat pack canisters are also optimally available with UN-Y approval for the storage of hazardous goods or with an integrated drain cock for safe emptying.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité