Vacuum system, EcoVac
Fournisseur: schuett-biotec GmbH
3350022
SCUT3350022EA 415 EUR InStock
SCUT3350022
Vacuum system, EcoVac
Pompe Systèmes de pompe à vide
The EcoVac safety-vacuum-system supports efficient work and increases the safety standard when working with infectious material in a laboratory.
  • The system easily removes pathogenic liquid excess from dishes, bottles, tubes and disks
  • Transfers to a safety vacuum bottle made of break-resistant and autoclavable polypropylene
The safety vacuum bottle is resistant to chemicals, has a large capacity and therefore decreases time-consuming autoclaving and disposing to a minimum. The screw-cap of the bottle comes with two self-locking fittings, which seal the bottle and lock the vacuum bottle hermetically, when the tubing is removed for transport and autoclaving.
The vacuum pump is a double chambered membrane pump with an illuminated on/off-switch. An optional foot switch allows to control operation of the pump, if EcoVac should be used in intermittently
Information de livraison: Includes safety PP-bottle (4 L) with screw cap, two self-locking fittings, aeration insert, safety filter and silicone tubing 5 m (autoclavable).
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité