Loop steriliser, Solaris
Fournisseur: schuett-biotec GmbH
3399002
SCUT3399002EA 1950 EUR InStock
SCUT3399002
Loop steriliser, Solaris
Stérilisateurs Stérilisateurs pour aiguilles
The Schuett Solaris is the perfect solution to avoid using naked flames or if gas cartridges are not available. The unit uses standard electrical power and reaches the safe working temperature of 900 to 1300 °C within seconds. No warm-up time, no excess heat, no energy consumption between work cycles. Start is generated by an infra-red sensor, which allows efficient one-handed operation. The integrated timer can be set to 5 or 7 s. Pathogenic material is kept inside the sterilisation chamber. No spreading, no sooting. Ideal for use in Laminar Flow Cabinets. Working angle can be adjusted. Robust and sturdy, no parts subject to mechanical wear, ideal for mobile operation in the field.
  • No gas, no naked flame
  • Instantly ready to use, no warm up time, no waiting
  • Touch free start, with IR-sensor and integrated timer
  • Economical, low-cost operation
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité