Bottle top dispenser, SEASTAR™ HF
Fournisseur: SEASTAR CHEMICALS
S077200-SSN003
SEACS077200-SSN003EA 0 EUR InStock
SEACS077200-SSN003
Bottle top dispenser, SEASTAR™ HF
Distributeurs pour flacons
SEASTAR™ HF bottle-top dispenser, constructed of fluoropolymer PFA or modified PTFE, combines the convenience of a traditional pump dispenser with the chemical compatibility and high purity afforded by fluoropolymers.
  • Can be used with high purity concentrated acid
  • Calibrated and mechanically tested to ensure each unit meets the required volume, precision and accuracy requirements
SEASTAR™ HF bottle-top dispenser is designed for applications which require the chemical resistance of fluoropolymers, such as ACS or technical grade acids (including hydrofluoric acid). These dispensers are calibrated and mechanically tested to ensure each unit meets the required volume, precision and accuracy requirements.
Materials: Body and plunger: PTFE TFM, valves and valve seats: PFA, handle and cap: HDPE and TFM, tubing and fittings: PFA.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité