Wiva™ VAT-5 Infinity Disposal Container - Polypropylene
Fournisseur: SEMADENI
12178
SEMA12178EA 23.6 EUR InStock
SEMA12178
Wiva™ VAT-5 Infinity Disposal Container - Polypropylene
Bidons de sécurité
Disposal container for clinical waste, chemical waste, and hazardous waste in laboratories.
  • Made from 100% recycled plastic
  • This reduces CO₂ emissions during production by 52%
The intelligent lid design offers a large number of advantages: Pressing gently on the lid causes the container to close, forming an odor-proof, airtight seal (day lid function); pressing firmly causes the container to hermetically seal for transport and disposal. The lid can be hung on the side of the container during use.
The click lid (item SEMA8687) has a round inner lid. Especially in places where the container is left standing for long periods of time and smaller amounts of waste are discarded, the round disposal opening can be conveniently opened and closed again. This significantly reduces the leakage of odor emissions. To lock the inner lid securely, this should be turned to the stop and then pressed down.
The containers are resistant to chemicals, heat and cold, and are very stable and drop-proof. They have two recessed and rounded handles.
Disposal containers are not suitable for transporting liquids; when they are securely closed for transport, the covers can no longer be opened. The containers are made from environmentally-friendly materials. Please observe the applicable local regulations for the disposal of hazardous waste. Cost effective purchase and transport costs; light and easy to handle; reduced space requirement because of square shape, stackable and palettable on top of each other.
Informations de commande: Without lid. Suitable lids must be ordered separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité