Wiva™ VAT-5 Infinity Disposal Container - Polypropylene
Fournisseur: SEMADENI
12178
SEMA12178EA 24.3 EUR InStock
SEMA12178
Wiva™ VAT-5 Infinity Disposal Container - Polypropylene
Bidons de sécurité
Disposal container for clinical waste, chemical waste, and hazardous waste in laboratories.
  • Made from 100% recycled plastic
  • This reduces CO₂ emissions during production by 52%
The intelligent lid design offers a large number of advantages: Pressing gently on the lid causes the container to close, forming an odor-proof, airtight seal (day lid function); pressing firmly causes the container to hermetically seal for transport and disposal. The lid can be hung on the side of the container during use.
The click lid (item SEMA8687) has a round inner lid.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité