Newbox, PP
Fournisseur: SEMADENI
12672
SEMA12672EA 0 EUR InStock
SEMA12672
Newbox, PP
Boîtes Boîtes de rangement
The closed Euro-standard container of the Newbox range boasts a slim design. Thanks to its robustness and dimensional stability, the Newbox is a durable and economic product.
  • Temperature resistant up to 80 °C (short term up to 100 °C)
  • 100% compatible with RAKO stacking containers
  • With two and four handle openings
The containers are stackable and dimensionally matched with Euro pallets. Compared with the Logiline range, the Newbox range impresses with a somewhat slimmer and lighter design that is nevertheless robust and dimensionally stable.
Applications: With manual handling closed Newbox containers are used universally as transport containers or for the storage of the widest possible range of goods.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité