Packaging bottles, wide neck, round, HDPE, with screw cap
Fournisseur: SEMADENI
5379
SEMA5379EA 587 EUR InStock
SEMA5379
Packaging bottles, wide neck, round, HDPE, with screw cap
Flacons Flacons en plastique
Fluorinated packaging bottles, HDPE, white, with screw cap.
  • Includes tamper-evident closure with resistant PET or aluminum gasket
  • Very high chemical resistance
  • With excellent fluorine treatment barrier properties
These bottles are fluorinated inwardly and are ideal for odour and flavorings, paints and varnishes, solvents and aggressive chemicals.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité