CSR Infinity H₂ Purifier
Fournisseur: SGE a Trajan company
202381-SS
SGEA202381-SSEA 555 EUR InStock
SGEA202381-SS
CSR Infinity H₂ Purifier
Filtres Filtres à gaz
The CRS Infinity H2 purifier is a self regenerating purifier that removes oxygen from hydrogen at ambient temperature.
  • High quality 316 stainless steel construction
  • For use at ambient temperatures
  • For use with hydrogen only
The reaction consumes the oxygen and creates an excess of water vapor. When paired with an additional moisture filter it is the perfect solution for hydrogen generators. This catalyic purifier has the potential to last forever, however the media can be easily contaminated by oil, organic solvents, heavy metals, sulfur compounds etc.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité