Biopsy bags, shandon
Fournisseur: Epredia
6774051 6774010 6774010W 6774019
SHAN6774051EA 477 EUR InStock
SHAN6774051 SHAN6774010 SHAN6774010W SHAN6774019
Biopsy bags, shandon
Sachets Sachets d'échantillonnage
These biopsy bags made of nylon, reduce reagent carry-over and pressure artifact often found with sponges.
  • Allow unrestricted fluid movement around tissue
  • Peel-open bag conveniently exposes specimen for rapid removal
These bags reduce the risk of small specimen loss. Acid resistant biopsy bags feature all the benefits of nylon biopsy bags, in addition acid-resistance for decalcification.
These biopsy bags are monofilament blue anti-static nylon that provide superior fluid dynamics and easy specimen identification due to the blue tint.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité