Electronic rotary microtome, HM 340 E
Fournisseur: Epredia
905190
SHAN905190EA 0 EUR InStock
SHAN905190
Electronic rotary microtome, HM 340 E
Microtomes et groupes de microtomes Microtomes
The HM 340 E is an ergonomically designed electronic rotary microtome. It features stepping-motor advance technology which provides superior sectioning results.
  • Selectable retraction
  • One-handed zero orientation head
  • Menu driven control panel that can be used on either side of the instrument
Designed for precision and stability, it yields quality ribbons even for difficult to cut sections. Applications range from paraffin sectioning in routine and research laboratories to plastic embedded specimens. The integrated waste tray captures excess paraffin debris to the rear of the blade holder for quick and easy cleaning. To guarantee safe specimen transfer the hand wheel brake locks in any position and the protective blade guard allows operation without exposing a blade edge.
Information de livraison: Supplied with 100 ml PARA/Gard™, standard tools, brushed aluminium plate cover, dust cover, and instruction manual.
Commander maintenant
COMMANDE
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité