Rotary microtome, HM 355 S
Fournisseur: Epredia
905200
SHAN905200EA 0 EUR InStock
SHAN905200
Rotary microtome, HM 355 S
Microtomes et groupes de microtomes Microtomes
The HM 355 S is a heavy duty microtome. User selected cutting window control maximises throughput by allowing the return stroke to be faster than the optimised cutting stroke.
  • Instant switch between trim and section settings with independently selectable trim and section speeds
  • Rock option: Specimen head advances by a set amount to trim specimens without turning the handwheel through full turns
  • Selectable retraction feature guide rail guidance
  • Emergency stop immediately cuts power to the motor
  • Viewing commands five languages are available
  • Display monitors functions including cutting speed, section count, section thickness and advance distance
Suitable for specially sophisticated paraffin and hard sectioning techniques in biology, medicine and industry.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité