Centrifuge, benchtop, Cytospin™ 4
Fournisseur: Epredia
A78300002
SHANA78300002EA 0 EUR InStock
SHANA78300002
Centrifuge, benchtop, Cytospin™ 4
Centrifugeuses Centrifugeuses de paillasse
This reliable benchtop centrifuge provides economical thin-layer preparations from any liquid matrix, especially hypocellular fluids such as spinal fluid and urine.
  • Processes 12 specimens at one time
  • Accepts all protocols from Cytospin 1, 2 and 3
  • Allows for one-handed opening and closing with a redesigned lid-release mechanism
  • Enables viewing of the sealed head through the polycarbonate window during operation
  • Allows loading of the sealed head inside a microbiological cabinet for ultimate safety
  • Features an auto-locking, plastic outer lid
  • Operates only when lid is locked and keeps lid locked during rotation
  • Includes disposable sample chambers with caps and a wipe-clean control panel
During operation, the instrument’s spinning action tilts the Shandon Cytofunnels® upright and centrifuges cells onto the deposition area of the slide, giving all cell types equal opportunity for presentation. In the Load or Stop position, this unique tilting feature reduces cell loss by preventing residual fluid from coming into contact with the prepared slide.
Information de livraison: Centrifuge is supplied with seated head/rotor, marking pen, operator's manual, service record book and standard workflow diagram.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité