Human Recombinant Leptin (Animal free) (from E. coli)
Fournisseur: Shenandoah Biotechnology
100-37AF-200UG
SHBT100-37AF-200UGEA 75.3 EUR InStock
SHBT100-37AF-200UG
Human Recombinant Leptin (Animal free) (from E. coli)
Protéines et Peptides
Leptin is a hormone that is produced by adipose tissue and plays critical roles in the physiologic regulation of body weight. Leptin acts through the leptin receptor (LEPR) to regulate adipose mass by inhibiting hunger and balancing energy usage. Leptin mutations cause severe hereditary obesity and hypogonadism in rodents and humans. Leptin also has thermogenic actions, regulates enzymes of fatty acid oxidation, and is involved in hematopoiesis, angiogenesis, wound healing, inflammation, and immune responses.
  • High Quality
  • Low Endotoxin
  • Affordable
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité