Human Recombinant FLT-3 (CTGrade GMP) (from E. coli)
Fournisseur: Shenandoah Biotechnology
500-03-100UG 500-03-1MG 500-03-50UG
SHBT500-03-100UGEA 2040 EUR InStock
SHBT500-03-100UG SHBT500-03-1MG SHBT500-03-50UG
Human Recombinant FLT-3 (CTGrade GMP) (from E. coli)
Protéines et Peptides
IL-6 is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages, and muscle cells. Signals through a receptor complex containing two receptors, IL-6Rα and gp130. Plays an important function in promoting fever and can serve to stimulate an immune response to trauma.
  • High quality
  • Low endotoxin
  • Lot-to-lot consistency
  • Animal free
  • Manufactured following GMP guidelines
IL-6 is often used for growth of hybridoma cell lines. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
Host cell protein: ≤30 ng/ml
Host cell DNA: ≤0,01 ng/ml
Sterility testing: No growth
Mycoplasma: Negative
Attention: For research use or further manufacturing only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité