Mouse Recombinant HSC Expansion PB (from E. coli)
Fournisseur: Shenandoah Biotechnology
PB-500-04
SHBTPB-500-04EA 251 EUR InStock
SHBTPB-500-04
Mouse Recombinant HSC Expansion PB (from E. coli)
Protéines et Peptides
Cytokines for polarizing T cells into T regulatory cells. T regulatory cells (Tregs) are a subset of T cells that suppress immune function. They are characterized by the expression of CD4, CD25 and Foxp3 and are known to produce immunosuppressive cytokines TGF-β 1 and IL-10. Tregs can be cultured from naive CD4+ T cells by culturing them in the presence of IL-2 and TGF-β 1 with CD3 and CD28 stimulation. In some cases cells are also cultured with neutralizing antibodies to IL-4, IFNγ and IL-12 and/or retinoic acid.
  • High quality
  • Low endotoxin
  • Affordable
Studies have shown that decreased expression of CD34 tends to correlate with decreased self renewal properties and pluripotent potential. Culturing CD34+ cells in a cytokine mixture of TPO, SCF, and FLT-3 Ligand for short time periods allows CD34+ cells to proliferate without decreasing CD34 expression, thus retaining their stem cell properties.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité