Mouse Th9 Cell Polarizing Recombinant Protein Bundle (rm IL-4, rh TGF-β 1) (from E. coli)
Fournisseur: Shenandoah Biotechnology
PB-500-16
SHBTPB-500-16EA 126 EUR InStock
SHBTPB-500-16
Mouse Th9 Cell Polarizing Recombinant Protein Bundle (rm IL-4, rh TGF-β 1) (from E. coli)
Protéines et Peptides
Cytokines for polarizing mouse T cells into Th9 cells. Th9 cells are a unique subset of T helper cells that produce IL-9. Although the exact function of IL-9 producing T cells is yet to be completely clear it is proposed that these cells could be important in inflammation and helminth infections. Th9 cells can be generated from naive CD4+ T cells in culture by culturing them in the presence of IL-4 and TGF-β 1. It also appears that IFNγ and IL-27 can inhibit the generation of Th9 cells.
  • High Quality
  • Low Endotoxin
  • Affordable
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité