Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Fournisseur: Shenandoah Biotechnology
PB-500-20
SHBTPB-500-20EA 835 EUR InStock
SHBTPB-500-20
Mouse Recombinant Mouse Glioma CultuPB (from E. coli)
Protéines et Peptides
Cytokines for cell culture of mouse glioma cells. A glioma is a generic term used to describe tumors that arise from the glia tissue of the brain that supports and surrounds neurons. Glioma tumor cells can be successfully cultured ex vivo with EGF, FGF-basic (or FGF-2), and PDGFs (AA and BB).
  • High quality
  • Low endotoxin
  • Affordable
Information de livraison: 2×100 μg of Recombinant mouse EGF, 200 μg of Recombinant mouse FGF-basic/FGF-2, 100 μg of Recombinant mouse PDGF-AA and 100 μg of Recombinant mouse PDGF-BB.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité