Temperature loggers, metal sheet and coil profiler
Fournisseur: Industrial Physics
CX3080
SIMECX3080EA 4587.45 EUR InStock
SIMECX3080
Temperature loggers, metal sheet and coil profiler
Enregistreurs de données
The metal sheet and coil coating cure profiler monitors the temperature profile of the coil strip at various points across its width as it is fed through the oven.
  • Made of aluminium/stainless steel
  • Six measuring points (3-air and 3-surface)
  • Easy grab handle
Without stopping the line, the metal sheet and coil coating cure profiler can be put on the moving coil. It is not possible to mount this system on vertical coil production facilities.
Information de livraison: Set includes special bracket, CurveX 2 USB oven logger, insulation box CX2003 (max.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité