Combotester HI 9813 Multi-Parameter Tester (pH/Conductivity/TDS/°C), Handheld
Fournisseur: Industrial Physics
HI9813
SIMEHI9813EA 294 EUR InStock
SIMEHI9813
Combotester HI 9813 Multi-Parameter Tester (pH/Conductivity/TDS/°C), Handheld
Electrochemistry Meters pH-mètres et multi-paramètres
The portable combo waterproof tester offer high accuracy pH, conductivity/TDS and temperature measurements in a single tester.
  • No more switching between meters for routine measurements
  • Waterproof, floating
  • Large, easy-to-read, dual-level LCD and automatic shut-off
  • pH and EC/TDS readings are automatically compensated for the effects of temperature (ATC)
  • EC/TDS conversion factor is user selectable as is the temperature compensation coefficient (ß)
This tester is supplied with a replaceable pH electrode cartridge with an extendable cloth junction as well as an EC/TDS graphite electrode that resists contamination by salts and other substances.
Information de livraison: Supplied with pH electrode, electrode removal tool, batteries (4 × 1,5 V) and instructions.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité