Reference guide and photographs for steel surfaces, SSPC-VIS 3
Fournisseur: Industrial Physics
LD3057
SIMELD3057EA 332 EUR InStock
SIMELD3057
Reference guide and photographs for steel surfaces, SSPC-VIS 3
Livres
SSPC-VIS 3 pictorial surface standard contains a series of 43 full-colour reference photographs to be used as a supplement to the SSPC standards for hand and power tool cleaned steel. Shows a total of seven different steel surfaces (four uncoated and three previously coated) before and after hand tool cleaning (SP 2), power tool cleaning with power wire brushes and sanding discs (SP 3), power tool cleaning to bare metal (SP 11), and commercial grade power tool cleaning (SP 15). It also contains pictures of SP 11 surfaces with a restored profile, a revised guide to the use of reference photographs, a new table of standards and conditions depicted and additional explanatory notes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité