Reference guide and photographs for steel surfaces, SSPC-VIS 5/NACE VIS 9
Fournisseur: Industrial Physics
LD3059
SIMELD3059EA 452 EUR InStock
SIMELD3059
Reference guide and photographs for steel surfaces, SSPC-VIS 5/NACE VIS 9
Livres
This pictorial standard shows two uncoated, rusted steel surfaces (Conditions C and D) before and after wet abrasive blast cleaning. Photographs illustrate two degrees of cleaning (WAB 6 and WAB 10) for each initial condition with additional photos that depict the appearance of light, moderate, and heavy flash rust after cleaning. Also contains a written guide to the use of reference photographs and additional explanatory notes. Cleanliness requirements for conditions depicted are defined by the joint SSPC/NACE surface preparation specifications for commercial (SP 6/NACE 3) and near-white blast cleaning (SP 10/NACE 2). Photos of commercial and near white surfaces achieved by dry abrasive blast cleaning can be found in SSPC-VIS 1.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité